Assay Details
Gene Symbol: Rgs8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250015.2 | XP_006250077.1 | 6 | 4539 | 83 | |
XM_006250016.2 | XP_006250078.1 | 7 | 4483 | 83 | ||
XM_006250017.2 | XP_006250079.1 | 7 | 4833 | 83 | ||
XM_008769675.1 | XP_008767897.1 | 7 | 4834 | 83 | ||
XM_008769676.1 | XP_008767898.1 | 7 | 4999 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
