Assay Details
Gene Symbol: Rgs8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250015.2 | XP_006250077.1 | 6 | 5530 | 107 | |
XM_006250016.2 | XP_006250078.1 | 7 | 5474 | 107 | ||
XM_006250017.2 | XP_006250079.1 | 7 | 5824 | 107 | ||
XM_008769675.1 | XP_008767897.1 | 7 | 5825 | 107 | ||
XM_008769676.1 | XP_008767898.1 | 7 | 5990 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
