Assay Details
Gene Symbol: Hsd11b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY387051.1 | - | 1905 | 104 |
Gene Symbol: Lrrgt00014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U83119.1 | - | 1 | 1896 | 104 |
Gene Symbol: Serinc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY539942.1 | - | 1302 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
