Assay Details
Gene Symbol: Crp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY310153.1 | - | 1345 | 136 | ||
AY318961.1 | - | 1345 | 136 | |||
AY321318.1 | - | 1342 | 136 | |||
AY321324.1 | - | 1345 | 136 | |||
AY325196.1 | - | 1345 | 136 | |||
AY325218.1 | - | 1345 | 136 | |||
AY325232.1 | - | 1345 | 136 |
Gene Symbol: LOC361942 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY539867.1 | - | 136 |
Gene Symbol: LOC499355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ100475.1 | - | 3 | 136 |
Gene Symbol: Vps13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY325251.1 | - | 10 | 1942 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



