Assay Details
Gene Symbol: Ttll11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001071863.5 | XP_001071863.3 | 1 | 114 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001071863.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
