Assay Details
Gene Symbol: Tnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236605.2 | XP_006236667.1 | 2 - 3 | 189 | 67 | |
XM_006236606.2 | XP_006236668.1 | 2 - 3 | 189 | 67 | ||
XM_008762999.1 | XP_008761221.1 | 2 - 3 | 189 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
