Assay Details
Gene Symbol: Suclg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100750.1 | NP_001094220.1 | 11 | 1604 | 111 | |
GenBank mRNA | BC079367.1 | - | 11 | 1598 | 111 | |
BC089863.1 | - | 10 | 1462 | 111 | ||
BC099746.1 | - | 11 | 1588 | 111 | ||
BC160886.1 | - | 11 | 1604 | 7385389 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
