Assay Details
Gene Symbol: Tspan9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107890.1 | NP_001101360.1 | 7 | 3141 | 111 | |
XM_006237494.2 | XP_006237556.1 | 11 | 3272 | 111 | ||
XM_006237495.2 | XP_006237557.1 | 10 | 3088 | 111 | ||
XM_006237496.2 | XP_006237558.1 | 10 | 3151 | 111 | ||
XM_006237498.2 | XP_006237560.1 | 9 | 2971 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
