Assay Details
Gene Symbol: Pacrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077677.1 | NP_001071145.1 | 1 | 221 | 106 | |
GenBank mRNA | BC127534.1 | - | 1 | 221 | 7133203 | 106 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC127534.1, NM_001077677.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
