Assay Details
Gene Symbol: Nhp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FQ217415.1 | - | 1018 | 125 | ||
FQ220734.1 | - | 1032 | 125 | |||
FQ233358.1 | - | 1032 | 125 |

More Information
Set Membership: |
![]() ![]() |
