Assay Details
Gene Symbol: Syt14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751296.3 | XP_003751344.1 | 3 - 4 | 444 | 72 | |
XM_003751297.3 | XP_003751345.1 | 3 - 4 | 496 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
