Assay Details
Gene Symbol: Syt14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751296.3 | XP_003751344.1 | 6 - 7 | 1384 | 72 | |
XM_003751297.3 | XP_003751345.1 | 6 - 7 | 1436 | 72 | ||
XM_008769895.1 | XP_008768117.1 | 5 - 6 | 2419 | 72 | ||
XM_008769896.1 | XP_008768118.1 | 5 - 6 | 2419 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
