Assay Details
Gene Symbol: Crp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY310153.1 | - | 626 | 106 | ||
AY318961.1 | - | 626 | 106 | |||
AY321318.1 | - | 623 | 106 | |||
AY321324.1 | - | 626 | 106 | |||
AY325196.1 | - | 626 | 106 | |||
AY325218.1 | - | 626 | 106 | |||
AY325232.1 | - | 626 | 106 |
Gene Symbol: LOC100233206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY539901.1 | - | 1423 | 106 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
