Assay Details
Gene Symbol: Poldip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130506.1 | NP_001123978.1 | 9 | 1931 | 73 | |
XM_006242090.2 | XP_006242152.1 | 8 | 1866 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
