Assay Details
Gene Symbol: Osbpl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_243980.7 | XP_243980.4 | 1 | 809 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_243980.7 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
