Assay Details
Gene Symbol: Tfeb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025707.1 | NP_001020878.1 | 8 | 2042 | 102 | |
XM_006244440.2 | XP_006244502.1 | 9 | 2971 | 102 | ||
XM_006244442.1 | XP_006244504.1 | 9 | 1961 | 102 | ||
XM_006244446.2 | XP_006244508.1 | 9 | 3232 | 102 | ||
GenBank mRNA | BC099102.1 | - | 8 | 2042 | 7460021 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
