Assay Details
Gene Symbol: Rab23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766955.1 | XP_008765177.1 | 1 | 345 | 100 | |
XM_008766956.1 | XP_008765178.1 | 1 | 345 | 100 | ||
XM_008766959.1 | XP_008765181.1 | 1 | 345 | 100 | ||
XM_008766960.1 | XP_008765182.1 | 1 | 345 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008766955.1, XM_008766956.1, XM_008766959.1, XM_008766960.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
