Assay Details
Gene Symbol: Dhcr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022389.2 | NP_071784.1 | 9 | 2345 | 85 | |
XM_006230892.2 | XP_006230954.1 | 8 | 2091 | 85 | ||
XM_006230893.2 | XP_006230955.1 | 8 | 1970 | 85 | ||
XM_006230894.2 | XP_006230956.1 | 9 | 1990 | 85 | ||
XM_008760170.1 | XP_008758392.1 | 9 | 3077 | 85 | ||
XM_008760171.1 | XP_008758393.1 | 8 | 2738 | 85 | ||
XM_008760172.1 | XP_008758394.1 | 9 | 2637 | 85 | ||
XM_008760173.1 | XP_008758395.1 | 8 | 2617 | 85 | ||
GenBank mRNA | AB016800.1 | - | 7 | 2035 | 85 | |
AF071500.1 | - | 8 | 1949 | 85 | ||
AF272393.1 | - | 9 | 2345 | 85 | ||
BC081688.1 | - | 7 | 2034 | 7109221 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
