Assay Details
Gene Symbol: Pdha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004072.2 | NP_001004072.2 | 11 | 2208 | 117 | |
GenBank mRNA | BC079369.1 | - | 11 | 2200 | 117 | |
BC098897.1 | - | 11 | 2208 | 7103543 | 117 | |
FQ214890.1 | - | 1 | 117 | |||
FQ235262.1 | - | 1 | 465 | 117 | ||
Z12158.1 | - | 11 | 2227 | 117 |
Gene Symbol: LOC103694482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773223.1 | XP_008771445.1 | 2 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
