Assay Details
Gene Symbol: Wrnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172332.2 | NP_758835.1 | 1 | 582 | 110 | |
GenBank mRNA | AY136827.1 | - | 1 | 486 | 110 | |
BC098652.1 | - | 1 | 582 | 7306780 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
