Assay Details
Gene Symbol: Sycp2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008771584.1 | XP_008769806.1 | 25 - 26 | 2428 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
