Assay Details
Gene Symbol: Samd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770515.1 | XP_008768737.1 | 11 | 5348 | 83 | |
XM_008770516.1 | XP_008768738.1 | 10 | 5083 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
