Assay Details
Gene Symbol: C1qtnf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191891.1 | NP_001178820.1 | 3 | 478 | 88 | |
XM_006252134.2 | XP_006252196.1 | 4 | 1258 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
