Assay Details
Gene Symbol: Prkaca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100922.1 | NP_001094392.1 | 10 | 1901 | 100 | |
XM_006255219.1 | XP_006255281.1 | 10 | 1834 | 100 | ||
GenBank mRNA | BC129128.1 | - | 10 | 1901 | 7314651 | 100 |
FQ213798.1 | - | 2 | 895 | 100 | ||
X57986.1 | - | 10 | 1808 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
