Assay Details
Gene Symbol: Anxa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013132.1 | NP_037264.1 | 13 | 1312 | 109 | |
XM_006232268.2 | XP_006232330.1 | 13 | 1449 | 109 | ||
GenBank mRNA | BC081855.1 | - | 13 | 1367 | 7192977 | 109 |
FQ213209.1 | - | 14 | 1444 | 109 | ||
FQ214176.1 | - | 13 | 1346 | 109 | ||
FQ229127.1 | - | 13 | 1302 | 109 | ||
M21730.1 | - | 13 | 1312 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
