Assay Details
Gene Symbol: Papss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106471.1 | NP_001099941.1 | 11 | 1891 | 77 | |
GenBank mRNA | BC166793.1 | - | 11 | 1964 | 7937610 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
