Assay Details
Gene Symbol: C1qtnf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012123.1 | NP_001012123.1 | 3 | 792 | 93 | |
GenBank mRNA | BC089992.1 | - | 3 | 792 | 7319641 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
