Assay Details
Gene Symbol: Pigq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007607.1 | NP_001007608.1 | 11 | 2385 | 102 | |
XM_006245942.2 | XP_006246004.1 | 11 | 2470 | 102 | ||
XM_008767559.1 | XP_008765781.1 | 11 | 2410 | 102 | ||
GenBank mRNA | BC081875.1 | - | 11 | 2385 | 7107681 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
