Assay Details
Gene Symbol: Mal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012798.1 | NP_036930.1 | 1 - 2 | 170 | 66 | |
GenBank mRNA | BC085755.1 | - | 1 - 2 | 131 | 66 | |
U31367.1 | - | 1 - 2 | 170 | 66 | ||
X82557.1 | - | 1 - 2 | 141 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
