Assay Details
Gene Symbol: Mal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012798.1 | NP_036930.1 | 2 - 3 | 339 | 95 | |
GenBank mRNA | BC085755.1 | - | 2 - 3 | 300 | 95 | |
U31367.1 | - | 2 - 3 | 339 | 95 | ||
X82557.1 | - | 2 - 3 | 310 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
