Assay Details
Gene Symbol: Icam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012967.1 | NP_037099.1 | 6 - 7 | 1491 | 99 | |
GenBank mRNA | BC081837.1 | - | 6 - 7 | 1503 | 99 | |
D00913.1 | - | 6 - 7 | 1491 | 99 | ||
FQ228241.1 | - | 6 - 7 | 1511 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
