Assay Details
Gene Symbol: Gnai1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013145.1 | NP_037277.1 | 6 - 7 | 939 | 66 | |
GenBank mRNA | DQ120469.1 | - | 6 - 7 | 813 | 66 | |
DQ120470.1 | - | 6 - 7 | 66 | |||
M17527.1 | - | 6 - 7 | 939 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
