Assay Details
Gene Symbol: Faf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017445.1 | NP_001017445.1 | 10 - 11 | 1041 | 82 | |
XM_006253617.2 | XP_006253679.1 | 9 - 10 | 1066 | 82 | ||
XM_008771488.1 | XP_008769710.1 | 7 - 8 | 850 | 82 | ||
GenBank mRNA | BC091224.1 | - | 10 - 11 | 1041 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
