Assay Details
Gene Symbol: Faf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017445.1 | NP_001017445.1 | 11 - 12 | 1187 | 79 | |
XM_006253617.2 | XP_006253679.1 | 10 - 11 | 1212 | 79 | ||
XM_008771488.1 | XP_008769710.1 | 8 - 9 | 996 | 79 | ||
GenBank mRNA | BC091224.1 | - | 11 - 12 | 1187 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
