Assay Details
Gene Symbol: Faf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017445.1 | NP_001017445.1 | 4 - 5 | 298 | 98 | |
XM_006253617.2 | XP_006253679.1 | 3 - 4 | 323 | 98 | ||
GenBank mRNA | BC079356.1 | - | 3 - 4 | 297 | 98 | |
BC091224.1 | - | 4 - 5 | 298 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC091224.1, NM_001017445.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
