Assay Details
Gene Symbol: Faf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017445.1 | NP_001017445.1 | 8 - 9 | 698 | 98 | |
XM_006253617.2 | XP_006253679.1 | 7 - 8 | 723 | 98 | ||
XM_008771488.1 | XP_008769710.1 | 5 - 6 | 507 | 98 | ||
GenBank mRNA | BC079356.1 | - | 7 - 8 | 697 | 98 | |
BC091224.1 | - | 8 - 9 | 698 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
