Assay Details
Gene Symbol: Il2ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013163.1 | NP_037295.1 | 7 - 8 | 897 | 80 | |
GenBank mRNA | FQ228242.1 | - | 929 | 80 | ||
M55049.1 | - | 7 - 8 | 897 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
