Assay Details
Gene Symbol: Cel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016997.2 | NP_058693.2 | 1 - 2 | 91 | 61 | |
GenBank mRNA | X16054.1 | - | 1 - 2 | 87 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
