Assay Details
Gene Symbol: Cel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016997.2 | NP_058693.2 | 5 - 6 | 695 | 57 | |
GenBank mRNA | X16054.1 | - | 5 - 6 | 691 | 57 | |
Z22803.1 | - | 4 - 5 | 614 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
