Assay Details
Gene Symbol: Cel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016997.2 | NP_058693.2 | 8 - 9 | 1109 | 70 | |
GenBank mRNA | M15893.1 | - | 8 - 9 | 1109 | 70 | |
X16054.1 | - | 8 - 9 | 1105 | 70 | ||
Z22803.1 | - | 7 - 8 | 1028 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
