Assay Details
Gene Symbol: Chrne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017194.1 | NP_058890.1 | 7 - 8 | 849 | 64 | |
XM_006246678.2 | XP_006246740.1 | 8 - 9 | 967 | 64 | ||
XM_008767807.1 | XP_008766029.1 | 8 - 9 | 950 | 64 | ||
XM_008767808.1 | XP_008766030.1 | 9 - 10 | 1616 | 64 | ||
GenBank mRNA | X13252.1 | - | 7 - 8 | 849 | 64 | |
X74836.1 | - | 7 - 8 | 849 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
