Assay Details
Gene Symbol: Gclm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017305.2 | NP_059001.1 | 6 - 7 | 771 | 93 | |
GenBank mRNA | BC078867.1 | - | 6 - 7 | 771 | 93 | |
DQ266366.1 | - | 5 - 6 | 513 | 93 | ||
L22191.1 | - | 6 - 7 | 709 | 93 | ||
S65555.1 | - | 6 - 7 | 709 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
