Assay Details
Gene Symbol: Rab27a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017317.2 | NP_059013.1 | 1 - 2 | 39 | 76 | |
GenBank mRNA | BC098667.1 | - | 1 - 2 | 39 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC098667.1, NM_017317.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
