Assay Details
Gene Symbol: Rab27a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017317.2 | NP_059013.1 | 2 - 3 | 220 | 76 | |
GenBank mRNA | BC098667.1 | - | 2 - 3 | 220 | 76 | |
D17352.1 | - | 1 - 2 | 156 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
