Assay Details
Gene Symbol: Fdft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019238.2 | NP_062111.1 | 3 - 4 | 459 | 98 | |
XM_008770746.1 | XP_008768968.1 | 2 - 3 | 3106 | 98 | ||
GenBank mRNA | BC081810.1 | - | 3 - 4 | 459 | 98 | |
FQ213323.1 | - | 3 - 4 | 442 | 98 | ||
FQ219372.1 | - | 3 - 4 | 428 | 98 | ||
M95591.1 | - | 3 - 4 | 477 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
