Assay Details
Gene Symbol: Fdft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019238.2 | NP_062111.1 | 4 - 5 | 577 | 84 | |
XM_008770746.1 | XP_008768968.1 | 3 - 4 | 3224 | 84 | ||
GenBank mRNA | BC081810.1 | - | 4 - 5 | 577 | 84 | |
FQ213323.1 | - | 4 - 5 | 560 | 84 | ||
FQ219372.1 | - | 4 - 5 | 546 | 84 | ||
M95591.1 | - | 4 - 5 | 595 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
