Assay Details
Gene Symbol: Fdft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019238.2 | NP_062111.1 | 5 - 6 | 783 | 83 | |
XM_008770746.1 | XP_008768968.1 | 4 - 5 | 3430 | 83 | ||
GenBank mRNA | BC081810.1 | - | 5 - 6 | 783 | 83 | |
M95591.1 | - | 5 - 6 | 801 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
