Assay Details
Gene Symbol: Cdk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019296.1 | NP_062169.1 | 1 - 2 | 71 | 79 | |
GenBank mRNA | FQ219810.1 | - | 1 - 2 | 74 | 81 | |
FQ227059.1 | - | 1 - 2 | 74 | 81 | ||
X60767.1 | - | 1 - 2 | 71 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_019296.1, X60767.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
