Assay Details
Gene Symbol: Cdk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019296.1 | NP_062169.1 | 4 - 5 | 411 | 62 | |
XM_006256353.2 | XP_006256415.1 | 5 - 6 | 538 | 62 | ||
GenBank mRNA | BC091549.1 | - | 5 - 6 | 496 | 62 | |
FQ219810.1 | - | 4 - 5 | 414 | 62 | ||
FQ227059.1 | - | 4 - 5 | 414 | 62 | ||
X60767.1 | - | 4 - 5 | 411 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
