Assay Details
Gene Symbol: Chrnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019297.1 | NP_062170.1 | 1 - 2 | 243 | 67 | |
GenBank mRNA | AY574258.1 | - | 1 - 2 | 55 | 67 | |
L31622.1 | - | 1 - 2 | 243 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
